Assay Details
Gene Symbol: Acadl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012819.1 | NP_036951.1 | 7 - 8 | 882 | 81 | |
GenBank mRNA | BC062006.1 | - | 7 - 8 | 880 | 5599828 | 81 |
J05029.1 | - | 7 - 8 | 893 | 81 | ||
L11276.1 | - | 7 - 8 | 882 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
