Assay Details
Gene Symbol: Anxa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012823.2 | NP_036955.2 | 2 - 3 | 149 | 108 | |
XM_006250694.2 | XP_006250756.1 | 2 - 3 | 513 | 108 | ||
XM_006250696.2 | XP_006250758.1 | 3 - 4 | 269 | 108 | ||
GenBank mRNA | BC081856.1 | - | 1 - 2 | 61 | 108 | |
FQ232970.1 | - | 2 - 3 | 140 | 108 | ||
M20559.1 | - | 2 - 3 | 97 | 108 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC081856.1, M20559.1, NM_012823.2, XM_006250694.2, XM_006250696.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
