Assay Details
Gene Symbol: Cacnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012828.2 | NP_036960.1 | 10 - 11 | 980 | 64 | |
XM_006257329.2 | XP_006257391.1 | 10 - 11 | 1052 | 64 | ||
XM_006257330.2 | XP_006257392.1 | 10 - 11 | 1461 | 64 | ||
GenBank mRNA | M88751.1 | - | 10 - 11 | 984 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
