Assay Details
Gene Symbol: Ephx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034090.3 | NP_001029262.1 | 6 - 7 | 1070 | 66 | |
NM_012844.3 | NP_036976.2 | 6 - 7 | 1099 | 66 | ||
GenBank mRNA | BC061568.1 | - | 6 - 7 | 1030 | 6889368 | 66 |
M26125.1 | - | 1075 | 66 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
liver development
cellular aromatic compound metabolic process
xenobiotic metabolic process
response to toxic substance
response to organic cyclic compound
aromatic compound catabolic process
cellular response to organic substance
cellular response to glucocorticoid stimulus
epoxide hydrolase activity
enzyme binding
cis-stilbene-oxide hydrolase activity
liver development
cellular aromatic compound metabolic process
xenobiotic metabolic process
response to toxic substance
response to organic cyclic compound
aromatic compound catabolic process
cellular response to organic substance
cellular response to glucocorticoid stimulus
epoxide hydrolase activity
enzyme binding
cis-stilbene-oxide hydrolase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P174374 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685126 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587838 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81820 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70845 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515533 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180703 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525095 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349843 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8430 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941629 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272866 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381488 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31593 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726244 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534310 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707064 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110453 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914926 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695845 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491131 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793940 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722846 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711086 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521771 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623845 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203368 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497880 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816572 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788318 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67535 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24135 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697314 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124356 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13274 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113983 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145407 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99514 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68568 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80393 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115452 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80950 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6858 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50816 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P174374 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P174374 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P174374 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P174374 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700589654 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 3041 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 68481 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 89998 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 36785 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 26386 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 27467 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 4738 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700589654 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10770348 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770350 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770345 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770343 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770344 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770346 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770347 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770349 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10770342 | - | - |
Rat Genome 230 2.0 Array | 1387669_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1387669_a_at | - | - |
Affymetrix HT Rat Focus Array | 1387669_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1387669_a_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1300014810.rn.1 | TC1300002080.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300014809.rn.1 | TC1300002080.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300014811.rn.1 | TC1300002080.rn.1 | RTA TC |
Rat Expression Set 230 | 1387669_a_at | - | - |
Rat Expression Set 230 | 1387669_PM_a_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770342 | - | - |
Rat Genome U34 Set | M26125_at | - | - |
Rat Genome U34 Set | M26125_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21567693 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1351186 | - | - |
RatRef-12 Expression BeadChip | ILMN_64601 | - | - |
