Assay Details
Gene Symbol: Fgf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012846.1 | NP_036978.1 | 1 - 2 | 228 | 64 | |
GenBank mRNA | X14232.1 | - | 1 - 2 | 228 | 64 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_012846.1, X14232.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
