Assay Details
Gene Symbol: Nfya | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012865.1 | NP_036997.1 | 5 - 6 | 611 | 94 | |
XM_006244409.2 | XP_006244471.1 | 5 - 6 | 618 | 94 | ||
XM_006244410.1 | XP_006244472.1 | 5 - 6 | 614 | 94 | ||
XM_006244411.2 | XP_006244473.1 | 5 - 6 | 608 | 94 | ||
XM_006244412.2 | XP_006244474.1 | 5 - 6 | 605 | 94 | ||
XM_006244413.1 | XP_006244475.1 | 4 - 5 | 526 | 94 | ||
XM_008766839.1 | XP_008765061.1 | 4 - 5 | 514 | 94 | ||
GenBank mRNA | FQ210330.1 | - | 4 - 5 | 531 | 94 | |
FQ223023.1 | - | 5 - 6 | 623 | 94 | ||
M34238.1 | - | 5 - 6 | 611 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
