Assay Details
Gene Symbol: Nfyc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012866.2 | NP_036998.1 | 6 - 7 | 758 | 57 | |
XM_006238785.2 | XP_006238847.1 | 6 - 7 | 731 | 57 | ||
GenBank mRNA | BC093619.1 | - | 6 - 7 | 758 | 57 | |
U17607.1 | - | 6 - 7 | 646 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
