Assay Details
Gene Symbol: Nfyc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012866.2 | NP_036998.1 | 9 - 10 | 1081 | 58 | |
XM_006238785.2 | XP_006238847.1 | 9 - 10 | 1054 | 58 | ||
GenBank mRNA | BC093619.1 | - | 9 - 10 | 1081 | 58 | |
U17607.1 | - | 9 - 10 | 969 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
transcription by RNA polymerase II
positive regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
transcription cis-regulatory region binding
RNA polymerase II cis-regulatory region sequence-specific DNA binding
DNA binding
DNA-binding transcription factor activity
transcription factor binding
sequence-specific DNA binding
protein heterodimerization activity
regulation of transcription, DNA-templated
transcription by RNA polymerase II
positive regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
transcription cis-regulatory region binding
RNA polymerase II cis-regulatory region sequence-specific DNA binding
DNA binding
DNA-binding transcription factor activity
transcription factor binding
sequence-specific DNA binding
protein heterodimerization activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P400520 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433780 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488280 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666564 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756673 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386596 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472657 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737372 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345691 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750765 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427478 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184037 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698419 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701189 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873691 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263926 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29274 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188490 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898095 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615597 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762386 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56701 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457444 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259113 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808485 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902595 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124027 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71798 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102031 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17091 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3114 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108838 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P400520 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P400520 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P400520 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P400520 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P174682 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700420281 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 99184 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 39721 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 67018 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 34642 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 25222 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 62026 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 73586 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 36883 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700420281 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10879428 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10879436 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10879434 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10879429 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10879433 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10879435 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10879439 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10879441 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10879438 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10879437 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10879431 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10879427 | - | - |
Rat Genome 230 2.0 Array | 1398268_PM_at | - | - |
Rat Genome 230 2.0 Array | 1368820_at | - | - |
Rat Genome 230 2.0 Array | 1368820_PM_at | - | - |
Rat Genome 230 2.0 Array | 1398268_at | - | - |
Affymetrix HT Rat Focus Array | 1368820_at | - | - |
Affymetrix HT Rat Focus Array | 1398268_at | - | - |
Affymetrix HT Rat Focus Array | 1368820_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1398268_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500026080.rn.1 | TC0500003522.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500026081.rn.1 | TC0500003522.rn.1 | RTA TC |
Rat Expression Set 230 | 1398268_PM_at | - | - |
Rat Expression Set 230 | 1368820_PM_at | - | - |
Rat Expression Set 230 | 1368820_at | - | - |
Rat Expression Set 230 | 1398268_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10879427 | - | - |
Rat Genome U34 Set | rc_AA875121_at | - | - |
Rat Genome U34 Set | U17607_at | - | - |
Rat Genome U34 Set | U17607_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358630 | - | - |
RatRef-12 Expression BeadChip | ILMN_64400 | - | - |
