Assay Details
Gene Symbol: Acadvl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012891.2 | NP_037023.1 | 7 - 8 | 642 | 69 | |
GenBank mRNA | BC087036.1 | - | 7 - 8 | 663 | 69 | |
D30647.1 | - | 7 - 8 | 644 | 69 | ||
FQ210754.1 | - | 7 - 8 | 691 | 69 | ||
FQ219329.1 | - | 7 - 8 | 687 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
