Assay Details
Gene Symbol: Adk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012895.3 | NP_037027.2 | 7 - 8 | 731 | 107 | |
XM_006251631.1 | XP_006251693.1 | 7 - 8 | 845 | 107 | ||
GenBank mRNA | BC081712.1 | - | 7 - 8 | 731 | 107 | |
FQ209450.1 | - | 2 - 3 | 200 | 107 | ||
FQ218369.1 | - | 3 - 4 | 394 | 107 | ||
FQ219573.1 | - | 1 - 2 | 39 | 107 | ||
FQ225093.1 | - | 3 - 4 | 402 | 107 | ||
U90340.1 | - | 7 - 8 | 734 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
