Assay Details
Gene Symbol: Casp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012922.2 | NP_037054.1 | 6 - 7 | 659 | 69 | |
XM_006253130.2 | XP_006253192.1 | 6 - 7 | 703 | 69 | ||
GenBank mRNA | BC081854.1 | - | 6 - 7 | 659 | 7104685 | 69 |
U49930.1 | - | 5 - 6 | 629 | 69 | ||
U84410.1 | - | 5 - 6 | 633 | 69 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
