Assay Details
Gene Symbol: Prkag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013010.2 | NP_037142.1 | 3 - 4 | 179 | 69 | |
XM_006257331.2 | XP_006257393.1 | 3 - 4 | 379 | 69 | ||
GenBank mRNA | BC085749.1 | - | 2 - 3 | 165 | 69 | |
BC097940.1 | - | 3 - 4 | 179 | 69 | ||
U42413.1 | - | 2 - 3 | 142 | 69 | ||
X95578.1 | - | 3 - 4 | 199 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
