Assay Details
Gene Symbol: Prkag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013010.2 | NP_037142.1 | 5 - 6 | 329 | 90 | |
XM_006257331.2 | XP_006257393.1 | 5 - 6 | 529 | 90 | ||
GenBank mRNA | BC085749.1 | - | 4 - 5 | 315 | 90 | |
BC097940.1 | - | 5 - 6 | 329 | 90 | ||
U42413.1 | - | 4 - 5 | 292 | 90 | ||
X95578.1 | - | 5 - 6 | 349 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
