Assay Details
Gene Symbol: Slc18a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013031.1 | NP_037163.1 | 8 - 9 | 942 | 83 | |
XM_006231664.2 | XP_006231726.1 | 8 - 9 | 1783 | 83 | ||
XM_008760515.1 | XP_008758737.1 | 7 - 8 | 1142 | 83 | ||
XM_008760516.1 | XP_008758738.1 | 8 - 9 | 1788 | 83 | ||
XM_008760517.1 | XP_008758739.1 | 8 - 9 | 1783 | 83 | ||
XM_008760518.1 | XP_008758740.1 | 7 - 8 | 1142 | 83 | ||
XM_008760519.1 | XP_008758741.1 | 6 - 7 | 1049 | 83 | ||
XM_008760520.1 | XP_008758742.1 | 5 - 6 | 965 | 83 | ||
GenBank mRNA | L00603.1 | - | 8 - 9 | 942 | 83 | |
M97381.1 | - | 8 - 9 | 913 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
