Assay Details
Gene Symbol: Slc6a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013034.4 | NP_037166.2 | 4 - 5 | 949 | 59 | |
XM_008767950.1 | XP_008766172.1 | 5 - 6 | 1005 | 59 | ||
GenBank mRNA | M79450.1 | - | 6 - 7 | 1073 | 59 | |
X63253.1 | - | 4 - 5 | 957 | 59 | ||
X63995.1 | - | 5 - 6 | 1002 | 59 | ||
Y11024.1 | - | 4 - 5 | 842 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
