Assay Details
Gene Symbol: Alpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013059.1 | NP_037191.1 | 11 - 12 | 1465 | 115 | |
XM_006239136.2 | XP_006239198.1 | 11 - 12 | 1839 | 115 | ||
GenBank mRNA | BC088399.1 | - | 11 - 12 | 1488 | 115 | |
J03572.1 | - | 11 - 12 | 1465 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
osteoblast differentiation
endochondral ossification
developmental process involved in reproduction
dephosphorylation
response to lipopolysaccharide
response to vitamin D
response to antibiotic
response to glucocorticoid
cellular response to organic cyclic compound
cementum mineralization
alkaline phosphatase activity
pyrophosphatase activity
metal ion binding
osteoblast differentiation
endochondral ossification
developmental process involved in reproduction
dephosphorylation
response to lipopolysaccharide
response to vitamin D
response to antibiotic
response to glucocorticoid
cellular response to organic cyclic compound
cementum mineralization
alkaline phosphatase activity
pyrophosphatase activity
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P11684 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712466 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533407 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969005 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917896 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705640 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502358 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755091 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589049 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809191 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881133 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920377 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75385 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545560 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115740 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660064 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633062 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689022 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211228 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185724 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706403 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588714 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264957 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696711 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27487 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47912 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56018 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52095 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P11684 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P11684 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P11684 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P11684 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 702149758 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 52511 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 4937 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 62273 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15367 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700817809 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 702149758 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10880814 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10880807 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10880806 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10880815 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10880816 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10880808 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10880817 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10880810 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10880809 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10880811 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10880812 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10880813 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10880805 | - | - |
Affymetrix Rat Clariom S Assay | PSR0500029247.rn.1 | TC0500003889.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500029246.rn.1 | TC0500003889.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500029245.rn.1 | TC0500003889.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10880805 | - | - |
Rat Genome U34 Set | J03572_i_at | - | - |
Rat Genome U34 Set | X16038exon_s_at | - | - |
Rat Genome U34 Set | rc_AI009183_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21537292 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1372113 | - | - |
RatRef-12 Expression BeadChip | ILMN_66199 | - | - |
