Assay Details
Gene Symbol: Alpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013059.1 | NP_037191.1 | 1 - 2 | 65 | 68 | |
GenBank mRNA | BC088399.1 | - | 1 - 2 | 88 | 68 | |
J03572.1 | - | 1 - 2 | 65 | 68 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC088399.1, J03572.1, NM_013059.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
