Assay Details
Gene Symbol: Alpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013059.1 | NP_037191.1 | 7 - 8 | 939 | 66 | |
XM_006239136.2 | XP_006239198.1 | 7 - 8 | 1313 | 66 | ||
GenBank mRNA | BC088399.1 | - | 7 - 8 | 962 | 66 | |
J03572.1 | - | 7 - 8 | 939 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
