Assay Details
Gene Symbol: Hspa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013083.2 | NP_037215.1 | 5 - 6 | 785 | 68 | |
GenBank mRNA | BC062017.1 | - | 5 - 6 | 745 | 68 | |
M14050.1 | - | 4 - 5 | 661 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
