Assay Details
Gene Symbol: Il15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013129.2 | NP_037261.1 | 3 - 4 | 236 | 140 | |
GenBank mRNA | AF015719.1 | - | 3 - 4 | 236 | 140 | |
U69272.1 | - | 781 | 140 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
