Assay Details
Gene Symbol: Ppard | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013141.2 | NP_037273.2 | 2 - 3 | 153 | 66 | |
XM_006256166.2 | XP_006256228.1 | 2 - 3 | 965 | 66 | ||
XM_008772797.1 | XP_008771019.1 | 1 - 2 | 1200 | 66 | ||
GenBank mRNA | FM033221.1 | - | 2 - 3 | 153 | 66 | |
U40064.1 | - | 2 - 3 | 175 | 66 | ||
U75918.1 | - | 2 - 3 | 167 | 66 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_013141.2, U40064.1, U75918.1, XM_006256166.2, XM_008772797.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
