Assay Details
Gene Symbol: Acadm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016986.2 | NP_058682.2 | 10 - 11 | 1054 | 84 | |
GenBank mRNA | FQ214755.1 | - | 4 - 5 | 382 | 84 | |
J02791.1 | - | 10 - 11 | 969 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
