Assay Details
Gene Symbol: Acadm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016986.2 | NP_058682.2 | 11 - 12 | 1299 | 96 | |
GenBank mRNA | FQ214755.1 | - | 5 - 6 | 627 | 96 | |
J02791.1 | - | 11 - 12 | 1214 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
liver development
glycogen biosynthetic process
organic acid metabolic process
regulation of gluconeogenesis
fatty acid beta-oxidation
heart development
response to nutrient
response to cold
response to xenobiotic stimulus
carnitine metabolic process
response to hormone
post-embryonic development
carnitine metabolic process, CoA-linked
fatty acid beta-oxidation using acyl-CoA dehydrogenase
response to starvation
carnitine biosynthetic process
response to copper ion
protein homotetramerization
response to glucocorticoid
medium-chain fatty acid metabolic process
medium-chain fatty acid catabolic process
cardiac muscle cell differentiation
fatty-acyl-CoA binding
acyl-CoA dehydrogenase activity
isomerase activity
identical protein binding
flavin adenine dinucleotide binding
medium-chain-acyl-CoA dehydrogenase activity
liver development
glycogen biosynthetic process
organic acid metabolic process
regulation of gluconeogenesis
fatty acid beta-oxidation
heart development
response to nutrient
response to cold
response to xenobiotic stimulus
carnitine metabolic process
response to hormone
post-embryonic development
carnitine metabolic process, CoA-linked
fatty acid beta-oxidation using acyl-CoA dehydrogenase
response to starvation
carnitine biosynthetic process
response to copper ion
protein homotetramerization
response to glucocorticoid
medium-chain fatty acid metabolic process
medium-chain fatty acid catabolic process
cardiac muscle cell differentiation
fatty-acyl-CoA binding
acyl-CoA dehydrogenase activity
isomerase activity
identical protein binding
flavin adenine dinucleotide binding
medium-chain-acyl-CoA dehydrogenase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P540774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949669 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511671 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497771 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673255 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751717 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191164 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409714 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653830 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469128 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423646 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634773 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325538 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140331 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164989 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69175 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P540774 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P540774 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P540774 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P540774 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701935714 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 702410484 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 80514 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 48275 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 26999 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 14916 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 702410484 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 701935714 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10827460 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827455 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827458 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827462 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827457 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827465 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827461 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827463 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827456 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827466 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827459 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827464 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10827454 | - | - |
Rat Genome 230 2.0 Array | 1367702_at | - | - |
Rat Genome 230 2.0 Array | 1367702_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1367702_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1367702_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0200034124.rn.1 | TC0200004932.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0200034123.rn.1 | TC0200004932.rn.1 | RTA TC |
Rat Expression Set 230 | 1367702_at | - | - |
Rat Expression Set 230 | 1367702_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827454 | - | - |
Rat Genome U34 Set | J02791_at | - | - |
Rat Genome U34 Set | J02791_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21286513 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1362897 | - | - |
RatRef-12 Expression BeadChip | ILMN_63879 | - | - |
