Assay Details
Gene Symbol: Acadm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016986.2 | NP_058682.2 | 1 - 2 | 140 | 88 | |
GenBank mRNA | CK359511.1 | - | 1 - 2 | 55 | 88 | |
J02791.1 | - | 1 - 2 | 55 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the J02791.1, NM_016986.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
