Assay Details
Gene Symbol: Acp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016988.2 | NP_058684.2 | 10 - 11 | 1176 | 69 | |
XM_006234500.1 | XP_006234562.1 | 11 - 12 | 2098 | 69 | ||
GenBank mRNA | BC081823.1 | - | 10 - 11 | 1147 | 69 | |
M27893.1 | - | 10 - 11 | 1145 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
