Assay Details
Gene Symbol: Epor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017002.2 | NP_058698.2 | 7 - 8 | 989 | 57 | |
GenBank mRNA | BC089810.1 | - | 7 - 8 | 989 | 7375416 | 57 |
D13566.1 | - | 7 - 8 | 907 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
