Assay Details
Gene Symbol: Pgm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017033.2 | NP_058729.2 | 10 - 11 | 1658 | 69 | |
XM_006238442.2 | XP_006238504.1 | 11 - 12 | 1849 | 69 | ||
XM_008763853.1 | XP_008762075.1 | 10 - 11 | 2201 | 69 | ||
GenBank mRNA | BC099807.1 | - | 10 - 11 | 1658 | 7937943 | 69 |
BC128703.1 | - | 10 - 11 | 1663 | 69 | ||
FQ220427.1 | - | 3 - 4 | 448 | 69 | ||
L11694.1 | - | 10 - 11 | 1641 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P498273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860500 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507196 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814334 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184178 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664233 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228561 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728888 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972963 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30810 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369747 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786219 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518690 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224055 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106366 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40948 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732810 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689868 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57606 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209292 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407016 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642467 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649829 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539560 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111858 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629768 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610874 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160358 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106659 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12985 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68105 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120391 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176859 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P498273 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P498273 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P498273 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P356078 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 66362 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 25509 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 9736 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 21849 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 74593 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10870184 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870185 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870179 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870189 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870181 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870188 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870186 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870180 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870183 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870182 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870178 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870187 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10870177 | - | - |
Rat Genome 230 2.0 Array | 1388634_at | - | - |
Rat Genome 230 2.0 Array | 1388634_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369473_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369473_at | - | - |
Affymetrix HT Rat Focus Array | 1369473_at | - | - |
Affymetrix HT Rat Focus Array | 1388634_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388634_at | - | - |
Affymetrix HT Rat Focus Array | 1369473_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500006753.rn.1 | TC0500001054.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500006752.rn.1 | TC0500001054.rn.1 | RTA TC |
Rat Expression Set 230 | 1388634_at | - | - |
Rat Expression Set 230 | 1388634_PM_at | - | - |
Rat Expression Set 230 | 1369473_PM_at | - | - |
Rat Expression Set 230 | 1369473_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870177 | - | - |
Rat Genome U34 Set | L11694_at | - | - |
Rat Genome U34 Set | U20195_s_at | - | - |
Rat Genome U34 Set | rc_AA894296_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20975314 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1372657 | - | - |
RatRef-12 Expression BeadChip | ILMN_56980 | - | - |
