Assay Details
Gene Symbol: Ppp3ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017041.1 | NP_058737.1 | 11 - 12 | 1448 | 77 | |
XM_006233363.2 | XP_006233425.1 | 11 - 12 | 1943 | 77 | ||
XM_006233364.2 | XP_006233426.1 | 11 - 12 | 1208 | 77 | ||
GenBank mRNA | D90035.1 | - | 11 - 12 | 1448 | 77 | |
FQ212355.1 | - | 11 - 12 | 1792 | 77 | ||
FQ212817.1 | - | 6 - 7 | 499 | 77 | ||
M29275.1 | - | 11 - 12 | 1448 | 77 | ||
X57115.1 | - | 11 - 12 | 1511 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
