Assay Details
Gene Symbol: Gabra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017069.3 | NP_058765.3 | 8 - 9 | 1164 | 121 | |
XM_006229511.2 | XP_006229573.1 | 8 - 9 | 1276 | 121 | ||
GenBank mRNA | CB725341.1 | - | 2 - 3 | 200 | 121 | |
X51991.1 | - | 8 - 9 | 1171 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
