Assay Details
Gene Symbol: Tyro3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017092.1 | NP_058788.1 | 4 - 5 | 574 | 90 | |
GenBank mRNA | D37880.1 | - | 4 - 5 | 574 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
