Assay Details
Gene Symbol: Akt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017093.1 | NP_058789.1 | 7 - 8 | 919 | 72 | |
XM_008759108.1 | XP_008757330.1 | 8 - 9 | 3318 | 72 | ||
XM_008759109.1 | XP_008757331.1 | 7 - 8 | 906 | 72 | ||
XM_008759110.1 | XP_008757332.1 | 7 - 8 | 3183 | 72 | ||
XM_008759111.1 | XP_008757333.1 | 7 - 8 | 1029 | 72 | ||
GenBank mRNA | D30041.1 | - | 7 - 8 | 919 | 72 | |
DQ198085.1 | - | 6 - 7 | 638 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of protein phosphorylation
glycogen biosynthetic process
glucose metabolic process
regulation of translation
protein phosphorylation
response to osmotic stress
multicellular organism development
insulin receptor signaling pathway
carbohydrate transport
positive regulation of signal transduction
positive regulation of gene expression
negative regulation of long-chain fatty acid import across plasma membrane
positive regulation of sodium ion transport
positive regulation of glucose metabolic process
response to muscle activity
positive regulation of cell migration
positive regulation of vesicle fusion
response to nutrient levels
positive regulation of fatty acid beta-oxidation
peripheral nervous system myelin maintenance
response to insulin
cellular response to insulin stimulus
cellular response to hormone stimulus
negative regulation of RNA splicing
positive regulation of peptidyl-serine phosphorylation
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
protein kinase B signaling
positive regulation of nitric oxide biosynthetic process
positive regulation of glycogen biosynthetic process
positive regulation of transcription by RNA polymerase II
positive regulation of glucose import
positive regulation of positive chemotaxis
intracellular protein transmembrane transport
cellular response to organic cyclic compound
protein localization to plasma membrane
positive regulation of protein targeting to membrane
positive regulation of cell motility
protein kinase activity
protein serine/threonine kinase activity
protein kinase C binding
protein binding
ATP binding
positive regulation of protein phosphorylation
glycogen biosynthetic process
glucose metabolic process
regulation of translation
protein phosphorylation
response to osmotic stress
multicellular organism development
insulin receptor signaling pathway
carbohydrate transport
positive regulation of signal transduction
positive regulation of gene expression
negative regulation of long-chain fatty acid import across plasma membrane
positive regulation of sodium ion transport
positive regulation of glucose metabolic process
response to muscle activity
positive regulation of cell migration
positive regulation of vesicle fusion
response to nutrient levels
positive regulation of fatty acid beta-oxidation
peripheral nervous system myelin maintenance
response to insulin
cellular response to insulin stimulus
cellular response to hormone stimulus
negative regulation of RNA splicing
positive regulation of peptidyl-serine phosphorylation
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
protein kinase B signaling
positive regulation of nitric oxide biosynthetic process
positive regulation of glycogen biosynthetic process
positive regulation of transcription by RNA polymerase II
positive regulation of glucose import
positive regulation of positive chemotaxis
intracellular protein transmembrane transport
cellular response to organic cyclic compound
protein localization to plasma membrane
positive regulation of protein targeting to membrane
positive regulation of cell motility
protein kinase activity
protein serine/threonine kinase activity
protein kinase C binding
protein binding
ATP binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P15267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348236 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398796 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629174 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971989 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265995 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745516 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350984 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488513 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81636 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686770 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600304 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346834 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222559 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34810 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916703 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727729 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446769 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350917 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893106 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766725 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798751 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381717 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224485 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108939 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139995 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143741 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102415 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P15267 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P15267 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P15267 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P15267 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700484422 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700330349 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 19846 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 5385 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 14486 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700484422 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700330349 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10705409 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10705405 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10705400 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10705410 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10705403 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10705397 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10705406 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10705401 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10705404 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10705398 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10705396 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10705408 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10705399 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10705402 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10705395 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10705407 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10705394 | - | - |
Rat Genome 230 2.0 Array | 1368832_at | - | - |
Rat Genome 230 2.0 Array | 1388765_PM_at | - | - |
Rat Genome 230 2.0 Array | 1388765_at | - | - |
Rat Genome 230 2.0 Array | 1387353_at | - | - |
Rat Genome 230 2.0 Array | 1368832_PM_at | - | - |
Rat Genome 230 2.0 Array | 1387353_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1387353_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368832_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388765_at | - | - |
Affymetrix HT Rat Focus Array | 1387353_at | - | - |
Affymetrix HT Rat Focus Array | 1388765_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368832_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100008314.rn.1 | TC0100001129.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100008313.rn.1 | TC0100001129.rn.1 | RTA TC |
Rat Expression Set 230 | 1387353_PM_at | - | - |
Rat Expression Set 230 | 1368832_at | - | - |
Rat Expression Set 230 | 1368832_PM_at | - | - |
Rat Expression Set 230 | 1387353_at | - | - |
Rat Expression Set 230 | 1388765_at | - | - |
Rat Expression Set 230 | 1388765_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10705394 | - | - |
Rat Genome U34 Set | rc_AI105076_s_at | - | - |
Rat Genome U34 Set | D30041_at | - | - |
Rat Genome U34 Set | rc_AA946508_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21335366 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1372084 | - | - |
RatRef-12 Expression BeadChip | ILMN_69821 | - | - |
