Assay Details
Gene Symbol: Mecr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017209.1 | NP_058905.1 | 3 - 4 | 424 | 81 | |
XM_006239009.2 | XP_006239071.1 | 4 - 5 | 543 | 81 | ||
GenBank mRNA | AB015724.1 | - | 3 - 4 | 424 | 81 | |
FQ210707.1 | - | 3 - 4 | 420 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
