Assay Details
Gene Symbol: Mecr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017209.1 | NP_058905.1 | 5 - 6 | 674 | 69 | |
XM_006239009.2 | XP_006239071.1 | 6 - 7 | 793 | 69 | ||
GenBank mRNA | AB015724.1 | - | 5 - 6 | 674 | 69 | |
FQ210707.1 | - | 5 - 6 | 670 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
