Assay Details
Gene Symbol: Slc3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017216.1 | NP_058912.1 | 6 - 7 | 1169 | 79 | |
GenBank mRNA | BC078852.1 | - | 6 - 7 | 1164 | 79 | |
M77345.1 | - | 6 - 7 | 1169 | 79 | ||
M80804.1 | - | 6 - 7 | 1195 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
