Assay Details
Gene Symbol: Uchl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017237.3 | NP_058933.2 | 2 - 3 | 99 | 81 | |
GenBank mRNA | BC060573.1 | - | 2 - 3 | 103 | 6920680 | 81 |
D10699.1 | - | 2 - 3 | 83 | 81 | ||
FQ214229.1 | - | 2 - 3 | 95 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
