Assay Details
Gene Symbol: Gabrd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017289.1 | NP_058985.1 | 5 - 6 | 602 | 92 | |
GenBank mRNA | BC087714.1 | - | 5 - 6 | 567 | 7317154 | 92 |
M35162.1 | - | 5 - 6 | 602 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
