Assay Details
Gene Symbol: Eci1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017306.4 | NP_059002.2 | 5 - 6 | 531 | 58 | |
GenBank mRNA | BC078705.1 | - | 5 - 6 | 531 | 58 | |
FQ213384.1 | - | 5 - 6 | 539 | 58 | ||
FQ218351.1 | - | 5 - 6 | 532 | 58 | ||
M61112.1 | - | 5 - 6 | 520 | 58 | ||
X61184.1 | - | 5 - 6 | 542 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
