Assay Details
Gene Symbol: Pdia3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017319.1 | NP_059015.1 | 7 - 8 | 914 | 94 | |
GenBank mRNA | BC062393.1 | - | 7 - 8 | 951 | 94 | |
D63378.1 | - | 7 - 8 | 914 | 94 | ||
FQ211627.1 | - | 1013 | 94 | |||
X12355.1 | - | 7 - 8 | 907 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
