Assay Details
Gene Symbol: Runx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017325.1 | NP_059021.1 | 4 - 5 | 1127 | 91 | |
XM_008768598.1 | XP_008766820.1 | 4 - 5 | 2929 | 91 | ||
GenBank mRNA | L35271.1 | - | 4 - 5 | 1127 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
