Assay Details
Gene Symbol: Tyms | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019179.1 | NP_062052.1 | 3 - 4 | 435 | 130 | |
XM_008767420.1 | XP_008765642.1 | 3 - 4 | 924 | 130 | ||
XM_008767421.1 | XP_008765643.1 | 4 - 5 | 480 | 130 | ||
GenBank mRNA | BC126093.1 | - | 3 - 4 | 449 | 130 | |
L12138.1 | - | 3 - 4 | 435 | 130 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
