Assay Details
Gene Symbol: Coq3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019187.1 | NP_062060.1 | 2 - 3 | 212 | 131 | |
GenBank mRNA | BC081811.1 | - | 377 | 131 | ||
BC098666.1 | - | 370 | 131 | |||
BC126094.1 | - | 384 | 131 | |||
L20427.1 | - | 2 - 3 | 212 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
