Assay Details
Gene Symbol: Aes | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019220.1 | NP_062093.1 | 5 - 6 | 458 | 95 | |
GenBank mRNA | BC101855.1 | - | 423 | 7933368 | 95 | |
L14462.1 | - | 5 - 6 | 458 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
