Assay Details
Gene Symbol: Srsf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019257.3 | NP_062130.2 | 119 | 58 | ||
GenBank mRNA | AF020683.1 | - | 1 - 2 | 181 | 58 | |
BC058479.1 | - | 113 | 58 | |||
FM081441.1 | - | 1 - 2 | 119 | 58 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF020683.1, BC058479.1, NM_019257.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
