Assay Details
Gene Symbol: Srsf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195505.1 | NP_001182434.1 | 814 | 131 | ||
NM_001195506.1 | NP_001182435.1 | 685 | 131 | |||
NM_019257.3 | NP_062130.2 | 689 | 131 | |||
GenBank mRNA | AF020683.1 | - | 6 - 7 | 1986 | 131 | |
BC058479.1 | - | 683 | 6920786 | 131 | ||
L13635.1 | - | 785 | 131 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
