Assay Details
Gene Symbol: Cd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019295.1 | NP_062168.1 | 3 - 4 | 483 | 87 | |
GenBank mRNA | BC098918.1 | - | 3 - 4 | 490 | 7384975 | 87 |
D10728.1 | - | 3 - 4 | 483 | 87 | ||
X78985.1 | - | 3 - 4 | 430 | 87 |
Gene Symbol: LOC100911215 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006231081.2 | XP_006231143.2 | 304 | 87 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
