Assay Details
Gene Symbol: Ppp2r2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022209.3 | NP_071545.2 | 7 - 8 | 1164 | 74 | |
NR_073588.1 | 10 - 11 | 1373 | 74 | |||
GenBank mRNA | AY251277.1 | - | 7 - 8 | 1116 | 74 | |
BC078834.1 | - | 10 - 11 | 1373 | 74 | ||
D38260.1 | - | 7 - 8 | 959 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
