Assay Details
Gene Symbol: Xiap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022231.2 | NP_071567.1 | 1 - 2 | 176 | 85 | |
GenBank mRNA | AB033366.1 | - | 1 - 2 | 175 | 85 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AB033366.1, NM_022231.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
