Assay Details
Gene Symbol: Ccnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053662.3 | NP_446114.1 | 764 | 143 | ||
XM_008761029.1 | XP_008759251.1 | 1840 | 143 | |||
GenBank mRNA | AF030091.3 | - | 764 | 143 | ||
BC085686.1 | - | 818 | 143 | |||
BC128699.1 | - | 858 | 143 | |||
BC161817.1 | - | 710 | 9022416 | 143 | ||
FQ213168.1 | - | 2 - 3 | 143 | |||
FQ232436.1 | - | 801 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
