Assay Details
Gene Symbol: Ccnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053662.3 | NP_446114.1 | 1224 | 75 | ||
XM_008761029.1 | XP_008759251.1 | 2300 | 75 | |||
GenBank mRNA | AF030091.3 | - | 1224 | 75 | ||
BC085686.1 | - | 1278 | 75 | |||
BC128699.1 | - | 1318 | 75 | |||
BC161817.1 | - | 1170 | 75 | |||
FQ213168.1 | - | 6 - 7 | 648 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
