Assay Details
Gene Symbol: Sept2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057148.2 | NP_476489.1 | 5 - 6 | 401 | 85 | |
XM_006245511.2 | XP_006245573.1 | 5 - 6 | 681 | 85 | ||
XM_006245512.2 | XP_006245574.1 | 5 - 6 | 684 | 85 | ||
XM_006245513.2 | XP_006245575.1 | 5 - 6 | 425 | 85 | ||
XM_006245514.1 | XP_006245576.1 | 5 - 6 | 459 | 85 | ||
GenBank mRNA | AB027561.1 | - | 4 - 5 | 362 | 85 | |
BC081745.1 | - | 5 - 6 | 401 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
